Anti-TGFB2 Mouse Monoclonal Antibody [clone: 220ct16.4.3.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031281EA 3700 DKK
USBI031281
Anti-TGFB2 Mouse Monoclonal Antibody [clone: 220ct16.4.3.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: TGFB2 (transforming growth factor, beta 2)
Clonality: Monoclonal
Clone: 220ct16.4.3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR