Anti-SWAP70 Mouse Monoclonal Antibody [clone: 434CT16.5.5]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031271EA 3700 DKK
USBI031271
Anti-SWAP70 Mouse Monoclonal Antibody [clone: 434CT16.5.5]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000

Type: Primary
Antigen: SWAP70 (SWAP switching B-cell complex 70kDa subunit)
Clonality: Monoclonal
Clone: 434CT16.5.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR