Anti-SPRED1 Mouse Monoclonal Antibody (Biotin) [clone: 462CT2.5.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031264-BIOTINEA 5310 DKK
USBI031264-BIOTIN
Anti-SPRED1 Mouse Monoclonal Antibody (Biotin) [clone: 462CT2.5.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: SPRED1 (sprouty-related, EVH1 domain containing 1)
Clonality: Monoclonal
Clone: 462CT2.5.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR