Anti-RPS6KA3 Mouse Monoclonal Antibody [clone: 356CT10.6.1.2]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031223EA 3940 DKK
USBI031223
Anti-RPS6KA3 Mouse Monoclonal Antibody [clone: 356CT10.6.1.2]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-16000

Type: Primary
Antigen: RPS6KA3 (ribosomal protein S6 kinase, 90kDa, polypeptide 3)
Clonality: Monoclonal
Clone: 356CT10.6.1.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR