Anti-Phosphatase, Alkaline, Placental Mouse Monoclonal Antibody (Biotin) [clone: 421CT8.1.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031194-BIOTINEA 5310 DKK
USBI031194-BIOTIN
Anti-Phosphatase, Alkaline, Placental Mouse Monoclonal Antibody (Biotin) [clone: 421CT8.1.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: Phosphatase, Alkaline, Placental
Clonality: Monoclonal
Clone: 421CT8.1.1
Conjugation: Biotin
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR