Anti-NUP50 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 247CT10.2.5]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI031170-PEEA 4600 DKK
USBI031170-PE
Anti-NUP50 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 247CT10.2.5]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:1560
Do not freeze! Light sensitive
Type: Primary
Antigen: NUP50 (nucleoporin 50kDa)
Clonality: Monoclonal
Clone: 247CT10.2.5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human