Anti-NUP50 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 247CT10.2.5]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI031170-APEA 4600 DKK
USBI031170-AP
Anti-NUP50 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 247CT10.2.5]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1560
Type: Primary
Antigen: NUP50 (nucleoporin 50kDa)
Clonality: Monoclonal
Clone: 247CT10.2.5
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human