Anti-Mobc Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: ID:19CT.2.8.14]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031133-HRPEA 4200 DKK
USBI031133-HRP
Anti-Mobc Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: ID:19CT.2.8.14]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: Mobc
Clonality: Monoclonal
Clone: ID:19CT.2.8.14
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR