Anti-METAP1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 248CT14.6.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031121-HRPEA 5310 DKK
USBI031121-HRP
Anti-METAP1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 248CT14.6.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: METAP1 (methionyl aminopeptidase 1)
Clonality: Monoclonal
Clone: 248CT14.6.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR