Anti-MET Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6AT44.279]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031119-PEEA 4200 DKK
USBI031119-PE
Anti-MET Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6AT44.279]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze! Light sensitive

Type: Primary
Antigen: MET (met proto-oncogene)
Clonality: Monoclonal
Clone: 6AT44.279
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR