Anti-MAP Kinase 15 Mouse Monoclonal Antibody [clone: 168CT10.6.6]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031100EA 3700 DKK
USBI031100
Anti-MAP Kinase 15 Mouse Monoclonal Antibody [clone: 168CT10.6.6]
Antistoffer
Recommended dilution: Western Blot: 1:1000; Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: MAP Kinase 15
Clonality: Monoclonal
Clone: 168CT10.6.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR