Anti-LOX Mouse Monoclonal Antibody [clone: 624CT23.7.3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031090EA 4380 DKK
USBI031090
Anti-LOX Mouse Monoclonal Antibody [clone: 624CT23.7.3]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: Monoclonal
Clone: 624CT23.7.3
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR