Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 534CT6.2.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031063-PEEA 5310 DKK
USBI031063-PE
Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 534CT6.2.1]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze! Light sensitive

Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: Monoclonal
Clone: 534CT6.2.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR