Anti-ITGAX/CD11c Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 428CT12.2.6.4]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031061-APCEA 6360 DKK
USBI031061-APC
Anti-ITGAX/CD11c Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 428CT12.2.6.4]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ITGAX/CD11c
Clonality: Monoclonal
Clone: 428CT12.2.6.4
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR