Anti-IGHD Mouse Monoclonal Antibody [clone: 607CT8.4.4]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031041EA 3940 DKK
USBI031041
Anti-IGHD Mouse Monoclonal Antibody [clone: 607CT8.4.4]
Antistoffer
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-800

Type: Primary
Antigen: IGHD (immunoglobulin heavy constant delta)
Clonality: Monoclonal
Clone: 607CT8.4.4
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR