Anti-HGF Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 489CT6.12.6]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031014-HRPEA 5310 DKK
USBI031014-HRP
Anti-HGF Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 489CT6.12.6]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: HGF (hepatocyte growth factor (hepapoietin A; scatter factor))
Clonality: Monoclonal
Clone: 489CT6.12.6
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR