Anti-Glutathione S Transferase Mouse Monoclonal Antibody [clone: 21CT54.13.1 / 21CT54.13.7]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI031004EA 3760 DKK
USBI031004
Anti-Glutathione S Transferase Mouse Monoclonal Antibody [clone: 21CT54.13.1 / 21CT54.13.7]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Tag
Antigen: Glutathione S Transferase
Clonality: Monoclonal
Clone: 21CT54.13.1 / 21CT54.13.7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR