Anti-GNPDA1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 237CT2.5.2]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030999-APEA 5310 DKK
USBI030999-AP
Anti-GNPDA1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 237CT2.5.2]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000

Type: Primary
Antigen: GNPDA1 (glucosamine-6-phosphate deaminase 1)
Clonality: Monoclonal
Clone: 237CT2.5.2
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR