Anti-GNAS Mouse Monoclonal Antibody [clone: 559CT 16.1.3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030997EA 4380 DKK
USBI030997
Anti-GNAS Mouse Monoclonal Antibody [clone: 559CT 16.1.3]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:300

Type: Primary
Antigen: GNAS (GNAS complex locus)
Clonality: Monoclonal
Clone: 559CT 16.1.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR