Anti-GFP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 168AT1211]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030991-APCEA 5440 DKK
USBI030991-APC
Anti-GFP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 168AT1211]
Antistoffer
Recommended dilution: FLISA: 1:3000; Western Blot: 1:2000; Immunochemistry: 1:50-100; Immunofluorescence: 1:50-100

Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: Monoclonal
Clone: 168AT1211
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR