Anti-MLST8 Mouse Monoclonal Antibody (Biotin) [clone: 211CT7.1.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030985-BIOTINEA 4200 DKK
USBI030985-BIOTIN
Anti-MLST8 Mouse Monoclonal Antibody (Biotin) [clone: 211CT7.1.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1560

Type: Primary
Antigen: MLST8 (MTOR associated protein, LST8 homolog (S. cerevisiae))
Clonality: Monoclonal
Clone: 211CT7.1.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR