Anti-GAB1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1AT1979]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030976-FITCEA 6360 DKK
USBI030976-FITC
Anti-GAB1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1AT1979]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:200-1000; Immunoprecipitation: 2-5 ?g

Type: Primary
Antigen: GAB1 (GRB2-associated binding protein 1)
Clonality: Monoclonal
Clone: 1AT1979
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR