Anti-FKRP Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 386CT13.1.9]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030965-APEA 5310 DKK
USBI030965-AP
Anti-FKRP Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 386CT13.1.9]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: FKRP (fukutin related protein)
Clonality: Monoclonal
Clone: 386CT13.1.9
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR