Anti-EZH2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 144CT2.1.1.5]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030954-HRPEA 5440 DKK
USBI030954-HRP
Anti-EZH2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 144CT2.1.1.5]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: EZH2 (enhancer of zeste homolog 2 (Drosophila))
Clonality: Monoclonal
Clone: 144CT2.1.1.5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR