Anti-ELAVL1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 224CT6.5.3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030942-APEA 5310 DKK
USBI030942-AP
Anti-ELAVL1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 224CT6.5.3]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ELAVL1 (ELAV like RNA binding protein 1)
Clonality: Monoclonal
Clone: 224CT6.5.3
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR