Anti-EGFR Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 688CT33.1.3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030930-HRPEA 5380 DKK
USBI030930-HRP
Anti-EGFR Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 688CT33.1.3]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000

Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: Monoclonal
Clone: 688CT33.1.3
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR