Anti-CTGF Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 558CT34.4.4]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030895-APEA 5310 DKK
USBI030895-AP
Anti-CTGF Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 558CT34.4.4]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: CTGF (connective tissue growth factor)
Clonality: Monoclonal
Clone: 558CT34.4.4
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR