Anti-CDC25C Mouse Monoclonal Antibody [clone: 233CT9.6.6]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBI030865EA 4380 DKK
USBI030865
Anti-CDC25C Mouse Monoclonal Antibody [clone: 233CT9.6.6]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-8000
Type: Primary
Antigen: CDC25C (cell division cycle 25C)
Clonality: Monoclonal
Clone: 233CT9.6.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human