Anti-CD55 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 561CT4.2.3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030856-PEEA 6360 DKK
USBI030856-PE
Anti-CD55 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 561CT4.2.3]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100
Do not freeze! Light sensitive

Type: Primary
Antigen: CD55 (CD55 molecule, decay accelerating factor for complement (Cromer blood group))
Clonality: Monoclonal
Clone: 561CT4.2.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR