Anti-CD34 Mouse Monoclonal Antibody (Biotin) [clone: 132CT29.1.5]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030848-BIOTINEA 5440 DKK
USBI030848-BIOTIN
Anti-CD34 Mouse Monoclonal Antibody (Biotin) [clone: 132CT29.1.5]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: Monoclonal
Clone: 132CT29.1.5
Conjugation: Biotin
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR