Anti-SDC1 Mouse Monoclonal Antibody [clone: 587CT7.3.6.5]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030840EA 4000 DKK
USBI030840
Anti-SDC1 Mouse Monoclonal Antibody [clone: 587CT7.3.6.5]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-8000; Flow Cytometry 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: SDC1 (syndecan 1)
Clonality: Monoclonal
Clone: 587CT7.3.6.5
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR