Anti-SDC1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 531CT15.4.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030837-APEA 5310 DKK
USBI030837-AP
Anti-SDC1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 531CT15.4.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: SDC1 (syndecan 1)
Clonality: Monoclonal
Clone: 531CT15.4.1;531CT15.1.4
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR