Anti-REL Mouse Monoclonal Antibody [clone: 318CT41.3.2]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030817EA 3940 DKK
USBI030817
Anti-REL Mouse Monoclonal Antibody [clone: 318CT41.3.2]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-16000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: REL (v-rel avian reticuloendotheliosis viral oncogene homolog)
Clonality: Monoclonal
Clone: 318CT41.3.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR