Anti-ATG7 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 29CT10.3.12]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030780-FITCEA 6360 DKK
USBI030780-FITC
Anti-ATG7 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 29CT10.3.12]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:200-1000

Type: Primary
Antigen: ATG7 (autophagy related 7)
Clonality: Monoclonal
Clone: 29CT10.3.12
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR