Anti-ANPEP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 496CT9.1.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030773-APCEA 5310 DKK
USBI030773-APC
Anti-ANPEP Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 496CT9.1.1]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: Monoclonal
Clone: 496CT9.1.1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR