Anti-Fetoprotein Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 143CT50.11.8]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030748-APEA 4620 DKK
USBI030748-AP
Anti-Fetoprotein Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 143CT50.11.8]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: Fetoprotein
Clonality: Monoclonal
Clone: 143CT50.11.8
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR