Anti-ADA Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 608CT2.1.3]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030746-FITCEA 5310 DKK
USBI030746-FITC
Anti-ADA Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 608CT2.1.3]
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: ADA (adenosine deaminase)
Clonality: Monoclonal
Clone: 608CT2.1.3
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR