Anti-ACVR1B Mouse Monoclonal Antibody (Biotin) [clone: 179CT11.2.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030744-BIOTINEA 4920 DKK
USBI030744-BIOTIN
Anti-ACVR1B Mouse Monoclonal Antibody (Biotin) [clone: 179CT11.2.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ACVR1B (activin A receptor, type IB)
Clonality: Monoclonal
Clone: 179CT11.2.1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR