Anti-ACTB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 137CT26.1.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030742-HRPEA 4620 DKK
USBI030742-HRP
Anti-ACTB Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 137CT26.1.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:200-4000; Immunochemistry1:50-100; Immunofluorescence: 1:100-500

Type: Primary
Antigen: ACTB (actin, beta)
Clonality: Monoclonal
Clone: 137CT26.1.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1+IgG1 Kappa
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR