Anti-ACTA1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 337CT30.10.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030740-HRPEA 4620 DKK
USBI030740-HRP
Anti-ACTA1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 337CT30.10.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Flow Cytometry 1:10-50

Type: Primary
Antigen: ACTA1 (actin, alpha 1, skeletal muscle)
Clonality: Monoclonal
Clone: 337CT30.10.1
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR