Anti-ACOX1 Rabbit Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 153CT43.1.1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030738-APEA 4620 DKK
USBI030738-AP
Anti-ACOX1 Rabbit Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 153CT43.1.1]
Antistoffer
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: ACOX1 (acyl-CoA oxidase 1, palmitoyl)
Clonality: Monoclonal
Clone: 153CT43.1.1
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR