Anti-WNT5A Mouse Monoclonal Antibody [clone: 13B803]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030725EA 3930 DKK
USBI030725
Anti-WNT5A Mouse Monoclonal Antibody [clone: 13B803]
Antistoffer
Type: Primary
Antigen: WNT5A (wingless-type MMTV integration site family, member 5A)
Clonality: Monoclonal
Clone: 13B803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR