Anti-UBE2I Mouse Monoclonal Antibody [clone: 13B791]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030713EA 3270 DKK
USBI030713
Anti-UBE2I Mouse Monoclonal Antibody [clone: 13B791]
Antistoffer
Type: Primary
Antigen: UBE2I (ubiquitin-conjugating enzyme E2I)
Clonality: Monoclonal
Clone: 13B791
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR