Anti-TYK2 Mouse Monoclonal Antibody [clone: 13B784]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030706EA 3330 DKK
USBI030706
Anti-TYK2 Mouse Monoclonal Antibody [clone: 13B784]
Antistoffer
Type: Primary
Antigen: TYK2 (tyrosine kinase 2)
Clonality: Monoclonal
Clone: 13B784
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR