Anti-TUBA8 Mouse Monoclonal Antibody [clone: 13B780]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030702EA 4410 DKK
USBI030702
Anti-TUBA8 Mouse Monoclonal Antibody [clone: 13B780]
Antistoffer
Type: Primary
Antigen: TUBA8 (tubulin, alpha 8)
Clonality: Monoclonal
Clone: 13B780
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR