Anti-TNNI2 Mouse Monoclonal Antibody [clone: 13B772]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030694EA 3270 DKK
USBI030694
Anti-TNNI2 Mouse Monoclonal Antibody [clone: 13B772]
Antistoffer
Type: Primary
Antigen: TNNI2 (troponin I type 2 (skeletal, fast))
Clonality: Monoclonal
Clone: 13B772
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR