Anti-TNK1 Mouse Monoclonal Antibody [clone: 13B771]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030693EA 3780 DKK
USBI030693
Anti-TNK1 Mouse Monoclonal Antibody [clone: 13B771]
Antistoffer
Type: Primary
Antigen: TNK1 (tyrosine kinase, non-receptor, 1)
Clonality: Monoclonal
Clone: 13B771
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR