Anti-THAP11 Mouse Monoclonal Antibody [clone: 13B762]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030684EA 3080 DKK
USBI030684
Anti-THAP11 Mouse Monoclonal Antibody [clone: 13B762]
Antistoffer
Type: Primary
Antigen: THAP11 (THAP domain containing 11)
Clonality: Monoclonal
Clone: 13B762
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR