Anti-TGFB1 Mouse Monoclonal Antibody [clone: 13B761]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030683EA 3270 DKK
USBI030683
Anti-TGFB1 Mouse Monoclonal Antibody [clone: 13B761]
Antistoffer
Type: Primary
Antigen: TGFB1 (transforming growth factor, beta 1)
Clonality: Monoclonal
Clone: 13B761
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR