Anti-SRC Mouse Monoclonal Antibody [clone: 13B735]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBI030657EA 3330 DKK
USBI030657
Anti-SRC Mouse Monoclonal Antibody [clone: 13B735]
Antistoffer
Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: Monoclonal
Clone: 13B735
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR